2'-Chloro-4'-(4-chlorophenoxy)acetophenone ≥98.0%

Supplier: TCI America

Synonyms: 1-[2-Chloro-4-(4-chlorophenoxy)phenyl]ethan-1-one4-Acetyl-3,4'-dichlorodiphenyl ether

C2219-25G
TCC2219-25GEA 227.92 USD
TCC2219-25G
2'-Chloro-4'-(4-chlorophenoxy)acetophenone ≥98.0%
2'-Chloro-4'-(4-chlorophenoxy)acetophenone

CAS Number: 119851-28-4
MDL Number: MFCD00140226
Molecular Formula: C14H10Cl2O2
Molecular Weight: 281.13
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 58


Other Product Names / Synonyms: 4-Acetyl-3,4'-dichlorodiphenyl Ether

Formula: C₁₄H₁₀Cl₂O₂
MW: 281.1340 g/mol
Melting Pt: 58 °C
MDL Number: MFCD00140226
CAS Number: 119851-28-4
https://us.vwr.com/stibo/web/std.lang.all/92/32/28939232.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR