5-Chloro-2-(methylthio)benzothiazole ≥98.0%

Supplier: TCI America

C2111-5G
TCC2111-5GEA 222 USD
TCC2111-5G
5-Chloro-2-(methylthio)benzothiazole ≥98.0%
5-Chloro-2-(methylthio)benzothiazole

CAS Number: 3507-41-3
Molecular Formula: C8H6ClNS2
Molecular Weight: 215.71
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 73

Formula: C₈H₆ClNS₂
Melting Pt: 73 °C
CAS Number: 3507-41-3
https://us.vwr.com/stibo/web/std.lang.all/91/62/28939162.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR