4'-Chloro-2',5'-dimethoxyacetoacetanilide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

C2100-25G
TCC2100-25GEA 58 USD
TCC2100-25G
4'-Chloro-2',5'-dimethoxyacetoacetanilide ≥98.0% (by HPLC, total nitrogen)
4'-Chloro-2',5'-dimethoxyacetoacetanilide

CAS Number: 4433-79-8
MDL Number: MFCD00026256
Molecular Formula: C12H14ClNO4
Molecular Weight: 271.70
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 108
Flash Point (°C): 170


Other Product Names / Synonyms: N-(4-Chloro-2,5-dimethoxyphenyl)acetoacetamide, Naphthol AS-IRG, Azoic Coupling Component 44

https://us.vwr.com/stibo/web/std.lang.all/91/55/28939155.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR