(1ɑ,2ɑ,4ɑ)-1,2,4-Cyclohexanetricarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2062-5G C2062-25G
TCC2062-5GEA 30.43 USD
TCC2062-5G TCC2062-25G
(1ɑ,2ɑ,4ɑ)-1,2,4-Cyclohexanetricarboxylic acid ≥98.0% (by GC, titration analysis)
(1alpha,2alpha,4alpha)-1,2,4-Cyclohexanetricarboxylic acid

CAS Number: 76784-95-7
Molecular Formula: C9H12O6
Molecular Weight: 216.19
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 182

Formula: C₉H₁₂O₆
MW: 216.19 g/mol
Melting Pt: 182 °C
CAS Number: 76784-95-7
https://us.vwr.com/stibo/web/std.lang.all/91/25/28939125.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR