3-Chloro-2,5-dimethylpyrazine ≥95.0%

Supplier: TCI America

C2040-5G
TCC2040-5GEA 114 USD
TCC2040-5G
3-Chloro-2,5-dimethylpyrazine ≥95.0%
3-Chloro-2,5-dimethylpyrazine

CAS Number: 95-89-6
Molecular Formula: C6H7ClN2
Molecular Weight: 142.59
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 114
Flash Point (°C): 83
Specific Gravity (20/20): 1.17

Formula: C₆H₇ClN₂
MW: 142.5900 g/mol
Boiling Pt: 114 °C
Density: 1.17 (20 °C)
Flash Pt: 83 °C
MDL Number: MFCD00006143
CAS Number: 95-89-6
https://us.vwr.com/stibo/web/std.lang.all/91/03/28939103.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR