1-(2-Cyanoethyl)-2-undecylimidazole ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
3-(2-Undecyl-1-imidazolyl)propionitrile
C1965-25G
TCC1965-25GEA
46.67
USD
TCC1965-25G
1-(2-Cyanoethyl)-2-undecylimidazole ≥98.0% (by titrimetric analysis)
1-(2-Cyanoethyl)-2-undecylimidazole
CAS Number: 23996-16-9
Molecular Formula: C17H29N3
Molecular Weight: 275.44
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 52
Flash Point (°C): 199
Other Product Names / Synonyms: 3-(2-Undecyl-1-imidazolyl)propionitrile
Formula:
C₁₇H₂₉N₃ MW: 275.44 g/mol Melting Pt: 52 °C Flash Pt: 199 °C |
CAS Number:
23996-16-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...