1-(2-Cyanoethyl)-2-methylimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-(2-Methyl-1H-imidazol-1-yl)propanenitrile3-(2-Methyl-1-imidazolyl)propionitrile

C1963-25G
TCC1963-25GEA 49.72 USD
TCC1963-25G
1-(2-Cyanoethyl)-2-methylimidazole ≥98.0% (by GC, titration analysis)
1-(2-Cyanoethyl)-2-methylimidazole

CAS Number: 23996-55-6
Molecular Formula: C7H9N3
Molecular Weight: 135.17
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 57
Flash Point (°C): 166


Other Product Names / Synonyms: 3-(2-Methyl-1-imidazolyl)propionitrile

Formula: C₇H₉N₃
MW: 135.17 g/mol
Melting Pt: 57 °C
Flash Pt: 166 °C
MDL Number: MFCD00047049
CAS Number: 23996-55-6
https://us.vwr.com/stibo/web/std.lang.all/90/37/28939037.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR