Coronene ≥98.0% (by HPLC), purified by sublimation

Supplier: TCI America

Synonyms: Hexabenzobenzene

C1961-100MG
TCC1961-100MGEA 181.69 USD
TCC1961-100MG
Coronene ≥98.0% (by HPLC), purified by sublimation
Coronene

CAS Number: 191-07-1
MDL Number: MFCD00004134
Molecular Formula: C24H12
Molecular Weight: 300.36
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Boiling point (°C): 525
Melting point (°C): 440


Product Categories: Materials Chemistry, Molecular Conductors, Nanocarbon Unit Structures (Nanocarbon Materials), Highly Purified Reagents
Other Product Names / Synonyms: Hexabenzobenzene

Formula: C₂₄H₁₂
MW: 300.36 g/mol
Boiling Pt: 525 °C
Melting Pt: 440 °C
Density: 1.371 g/cm³ (25 °C)
MDL Number: MFCD00004134
CAS Number: 191-07-1
EINECS: 205-881-7
https://us.vwr.com/stibo/web/std.lang.all/90/35/28939035.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR