N-(4-Chlorophenyl)formamide ≥98.0%
Supplier: TCI America
Synonyms:
4'-Chloroformanilide
, 1-Chloro-4-formamidobenzene
C1945-5G
TCC1945-5GEA
156
USD
TCC1945-5G
N-(4-Chlorophenyl)formamide ≥98.0%
N-(4-Chlorophenyl)formamide
CAS Number: 2617-79-0
MDL Number: MFCD00021020
Molecular Formula: C7H6ClNO
Molecular Weight: 155.58
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 104
Other Product Names / Synonyms: 1-Chloro-4-formamidobenzene, 4'-Chloroformanilide
Formula:
C₇H₆ClNO MW: 155.58 g/mol Melting Pt: 104 °C |
MDL Number:
MFCD00021020 CAS Number: 2617-79-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...