15-Crown-4 [4-(2,4-dinitrophenylazo)phenol] ≥98.0% (by HPLC)

Synonyms: 5-(2,4-Dinitrophenylazo)-2-hydroxy-1,3-xylylene-15-crown 4-Ether

C1942-100MG
TCC1942-100MGEA 276.42 USD
TCC1942-100MG
15-Crown-4 [4-(2,4-dinitrophenylazo)phenol] ≥98.0% (by HPLC)
15-Crown-4 [4-(2,4-dinitrophenylazo)phenol]

CAS Number: 81238-57-5
Molecular Formula: C20H22N4O9
Molecular Weight: 462.42
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 5-(2,4-Dinitrophenylazo)-2-hydroxy-1,3-xylylene-15-crown 4-Ether

Formula: C₂₀H₂₂N₄O₉
MW: 462.42 g/mol
CAS Number: 81238-57-5
https://us.vwr.com/stibo/web/std.lang.all/90/19/28939019.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR