N-(2-Carboxy-4,5-dichlorobenzoyl)-(+)-10,2-camphorsultam ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C1767-500MG
TCC1767-500MGEA 325 USD
TCC1767-500MG
N-(2-Carboxy-4,5-dichlorobenzoyl)-(+)-10,2-camphorsultam ≥98.0% (by HPLC, titration analysis)
N-(2-Carboxy-4,5-dichlorobenzoyl)-(+)-10,2-camphorsultam

MDL Number: MFCD04117916
Molecular Formula: C18H19Cl2NO5S
Molecular Weight: 432.31
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 208
Specific rotation [a]20/D: 95 deg (C=1, MeOH)

https://us.vwr.com/stibo/web/std.lang.all/89/57/28938957.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR