N-(2-Carboxybenzoyl)-(+)-10,2-camphorsultam ≥98.0% (by HPLC)
Supplier: TCI America
C1766-500MG
TCC1766-500MGEA
339.87
USD
TCC1766-500MG
N-(2-Carboxybenzoyl)-(+)-10,2-camphorsultam ≥98.0% (by HPLC)
N-(2-Carboxybenzoyl)-(+)-10,2-camphorsultam
MDL Number: MFCD04117915
Molecular Formula: C18H21NO5S
Molecular Weight: 363.43
Purity/Analysis Method:
Form: Crystal
Specific rotation [a]20/D: 135 deg (C=2, MeOH)
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...