3-Chloro-2,4,5,6-tetrafluorobenzotrifluoride ≥98.0%
Supplier: TCI America
Synonyms:
3-Chloro-alpha,alpha,alpha,2,4,5,6-heptafluorotoluene
CAS Number: 4284-09-7
MDL Number: MFCD03094143
Molecular Formula: C7ClF7
Molecular Weight: 252.52
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 137
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Chloro-alpha,alpha,alpha,2,4,5,6-heptafluorotoluene
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...