4'-Carboxybenzo-15-crown-5 (4'-carboxybenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,3,5,6,8,9,11,12-Octahydrobenzo[b][1,4,7,10,13]pentaoxacyclopentadecine-15-carboxylic acid4'-Carboxybenzo-15-crown 5-Ether(Benzo-15-crown 5-Ether)-4'-carboxylic acid2,3-(4-Carboxybenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene

C1713-1G
TCC1713-1GEA 653 USD
TCC1713-1G
4'-Carboxybenzo-15-crown-5 (4'-carboxybenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥98.0% (by GC, titration analysis)
4'-Carboxybenzo-15-crown-5 (4'-Carboxybenzo-1,4,7,10,13-pentaoxacyclopentadecane)

CAS Number: 56683-55-7
MDL Number: MFCD00143276
Molecular Formula: C15H20O7
Molecular Weight: 312.32
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 189


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: (Benzo-15-crown 5-Ether)-4'-carboxylic Acid, 2,3-(4-Carboxybenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene

https://us.vwr.com/stibo/web/std.lang.all/89/16/28938916.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR