6-Chloro-3-formyl-7-methylchromone ≥95.0% (by titrimetric analysis)

C1694-5G
TCC1694-5GEA 97.8 USD
TCC1694-5G
6-Chloro-3-formyl-7-methylchromone ≥95.0% (by titrimetric analysis)
6-Chloro-3-formyl-7-methylchromone

CAS Number: 64481-12-5
MDL Number: MFCD00191919
Molecular Formula: C11H7ClO3
Molecular Weight: 222.62
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Melting point (°C): 182


Other Product Names / Synonyms: 6-Chloro-7-methyl-4-oxo-4H-1-benzopyran-3-carboxaldehyde, 6-Chloro-7-methylchromone-3-carboxaldehyde

Formula: C₁₁H₇ClO₃
Melting Pt: 182 °C
MDL Number: MFCD00191919
CAS Number: 64481-12-5
https://us.vwr.com/stibo/web/std.lang.all/89/05/28938905.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR