4-(2-Chloroethyl)anisole ≥96.0%
Supplier: TCI America
Synonyms:
1-(2-Chloroethyl)-4-methoxybenzene
C1680-25G
TCC1680-25GEA
103.53
USD
TCC1680-25G
4-(2-Chloroethyl)anisole ≥96.0%
4-(2-Chloroethyl)anisole
CAS Number: 18217-00-0
MDL Number: MFCD00044718
Molecular Formula: C9H11ClO
Molecular Weight: 170.64
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 139
Flash Point (°C): 122
Specific Gravity (20/20): 1.12
Other Product Names / Synonyms: 1-(2-Chloroethyl)-4-methoxybenzene
Formula:
C₉H₁₁ClO Boiling Pt: 139 °C Density: 1.12 (20 °C) Flash Pt: 122 °C |
MDL Number:
MFCD00044718 CAS Number: 18217-00-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...