2-(2-Chloroethoxy)tetrahydropyran ≥96.0%
Supplier: TCI America
Synonyms:
2-(2-Chloroethoxy)tetrahydro-2H-pyran
C1678-25G
TCC1678-25GEA
165
USD
TCC1678-25G
2-(2-Chloroethoxy)tetrahydropyran ≥96.0%
2-(2-Chloroethoxy)tetrahydropyran
CAS Number: 5631-96-9
MDL Number: MFCD00142761
Molecular Formula: C7H13ClO2
Molecular Weight: 164.63
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): 65
Specific Gravity (20/20): 1.12
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...