2-Chlorocyclododecanone ≥97.0%

Supplier: TCI America

C1650-5G C1650-1G
TCC1650-5GEA 472 USD
TCC1650-5G TCC1650-1G
2-Chlorocyclododecanone ≥97.0%
2-Chlorocyclododecanone

CAS Number: 35951-28-1
MDL Number: MFCD04117912
Molecular Formula: C12H21ClO
Molecular Weight: 216.75
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Storage Temperature: 0-10°C

Formula: C₁₂H₂₁ClO
MW: 216.75 g/mol
MDL Number: MFCD04117912
CAS Number: 35951-28-1
https://us.vwr.com/stibo/web/std.lang.all/88/64/28938864.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR