5-Chloro-1-methyl-4-nitroimidazole ≥98.0% (by total nitrogen basis)

Supplier: TCI America

C1646-5G
TCC1646-005GEA 101 USD
TCC1646-005G
5-Chloro-1-methyl-4-nitroimidazole ≥98.0% (by total nitrogen basis)
5-Chloro-1-methyl-4-nitroimidazole

CAS Number: 4897-25-0
MDL Number: MFCD00233664
Molecular Formula: C4H4ClN3O2
Molecular Weight: 161.55
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Melting point (°C): 147

Formula: C₄H₄ClN₃O₂
MW: 161.55 g/mol
Melting Pt: 147 °C
MDL Number: MFCD00233664
CAS Number: 4897-25-0
https://us.vwr.com/stibo/web/std.lang.all/88/61/28938861.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR