1-Chloromethylpyrene ≥97.0% (by titrimetric analysis)
Supplier: TCI America
C1637-1G
TCC1637-001GEA
406
USD
TCC1637-001G
1-Chloromethylpyrene ≥97.0% (by titrimetric analysis)
1-Chloromethylpyrene
CAS Number: 1086-00-6
MDL Number: MFCD00962113
Molecular Formula: C17H11Cl
Molecular Weight: 250.73
Purity/Analysis Method:
Form: Crystal
Storage Temperature:
Formula:
C₁₇H₁₁Cl |
MDL Number:
MFCD00962113 CAS Number: 1086-00-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...