(-)-B-Chlorodiisopinocampheylborane 60% (∼1.7 mol/L) in hexane

Danger

C1615-100ML
TCC1615-100MLEA 199.92 USD
TCC1615-100ML
(-)-B-Chlorodiisopinocampheylborane 60% (∼1.7 mol/L) in hexane
(-)-B-Chlorodiisopinocampheylborane

Caution: Store under inert gas

Formula: C₂₀H₃₄BCl
Boiling Pt: 69 °C
Density: 0.88 (20°C)
Storage Temperature: Refrigerator
MDL Number: MFCD00074807
CAS Number: 85116-37-6
UN: 2924
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/88/33/28938833.jpg

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (Argentmetric titration) 60.0 to 66.0 w/w%
Specific rotation [a]20/D -47.0 to -54.0 deg (neat) (Calcd. on Hexane solution)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR