(+)-B-Chlorodiisopinocampheylborane 58% ∼1.6 mol/L in hexane

Supplier: TCI America

Synonyms: (+)-Diisopinocampheylchloroborane

C1614-100ML
TCC1614-100MLEA 272.02 USD
TCC1614-100ML
(+)-B-Chlorodiisopinocampheylborane 58% ∼1.6 mol/L in hexane
(+)-B-Chlorodiisopinocampheylborane

CAS Number: 112246-73-8
MDL Number: MFCD00074808
Molecular Formula: C20H34BCl
Molecular Weight: 320.75
Form: Clear Liquid
Color: Colorless
Flash Point (°C): -18
Specific Gravity (20/20): 0.88
Specific rotation [a]20/D: 51 deg (neat)
Storage Temperature: 0-10°C

Formula: C₂₀H₃₄BCl
MW: 320.75 g/mol
Density: 0.88 (20 °C)
Flash Pt: –18 °C
MDL Number: MFCD00074808
CAS Number: 112246-73-8
https://us.vwr.com/stibo/web/std.lang.all/88/32/28938832.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR