1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine hydrochloride ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

C1608-25G
TCC1608-025GEA 183.65 USD
TCC1608-025G
1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine hydrochloride ≥97.0% (by HPLC, titration analysis)
1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine hydrochloride

CAS Number: 52605-52-4
MDL Number: MFCD00067039
Molecular Formula: C13H18Cl2N2
Molecular Weight: 309.66
Purity/Analysis Method: 97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 201

Formula: C₁₃H₁₈Cl₂N₂
Melting Pt: 201 °C
MDL Number: MFCD00067039
CAS Number: 52605-52-4
https://us.vwr.com/stibo/web/std.lang.all/88/25/28938825.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR