Chloromethyl-p-tolyl sulfide ≥96.0%
Supplier: TCI America
Synonyms:
4-(Chloromethylthio)toluene
C1601-5G
TCC1601-005GEA
209
USD
TCC1601-005G
Chloromethyl-p-tolyl sulfide ≥96.0%
Chloromethyl-p-tolyl sulfide
CAS Number: 34125-84-3
MDL Number: MFCD01318451
Molecular Formula: C8H9ClS
Molecular Weight: 172.67
Purity/Analysis Method:
Form: Clear Liquid
Specific Gravity (20/20): 1.17
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Sulfides
Other Product Names / Synonyms: 4-(Chloromethylthio)toluene, p-Tolyl Chloromethyl Sulfide
Formula:
C₈H₉ClS MW: 172.67 g/mol Density: 1.17 (20 °C) |
MDL Number:
MFCD01318451 CAS Number: 34125-84-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...