Bis(cyclopentadienyl)cobalt(III) hexafluorophosphate ≥96.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Cobaltocene Hexafluorophosphate
, Cobalticinium hexafluorophosphate
C1589-1G
TCC1589-001GEA
209.18
USD
TCC1589-001G
Bis(cyclopentadienyl)cobalt(III) hexafluorophosphate ≥96.0% (by titrimetric analysis)
Bis(cyclopentadienyl)cobalt(III) hexafluorophosphate
CAS Number: 12427-42-8
MDL Number: MFCD00011874
Molecular Formula: C10H10CoF6P
Molecular Weight: 334.09
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: Cobaltocene Hexafluorophosphate
Formula:
C₁₀H₁₀CoF₆P MW: 334.09 g/mol |
MDL Number:
MFCD00011874 CAS Number: 12427-42-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...