Cycloheptanone oxime ≥98.0%
Supplier: TCI America
Synonyms:
Suberoxime
C1543-25G
TCC1543-025GEA
265.81
USD
TCC1543-025G
Cycloheptanone oxime ≥98.0%
Cycloheptanone oxime
CAS Number: 2158-31-8
MDL Number: MFCD00046508
Molecular Formula: C7H13NO
Molecular Weight: 127.19
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 152
Specific Gravity (20/20): 1.03
Other Product Names / Synonyms: Suberoxime
Formula:
C₇H₁₃NO MW: 127.19 g/mol Boiling Pt: 152 °C Density: 1.03 (20 °C) |
MDL Number:
MFCD00046508 CAS Number: 2158-31-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...