Crocin (gardenia fruits extract)

Supplier: TCI America

Synonyms: Crocetin Digentiobiosyl Ester

C1527-25G
TCC1527-025GEA 186.76 USD
TCC1527-025G
Crocin (gardenia fruits extract)
Crocin (gardenia fruits extract)

CAS Number: 42553-65-1
MDL Number: MFCD00017495
Molecular Formula: C44H64O24
Molecular Weight: 976.97
Form: Crystal
Color: Yellow


Other Product Names / Synonyms: Crocetin Digentiobiosyl Ester

Formula: C₄₄H₆₄O₂₄
MW: 976.97 g/mol
MDL Number: MFCD00017495
CAS Number: 42553-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR