1-(Chloromethyl)benzotriazole ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1-(Chloromethyl)-1H-benzotriazole

C1526-25G C1526-5G
TCC1526-005GEA 98.46 USD
TCC1526-005G TCC1526-025G
1-(Chloromethyl)benzotriazole ≥98.0% (by titrimetric analysis)
1-(Chloromethyl)benzotriazole

CAS Number: 54187-96-1
MDL Number: MFCD00216649
Molecular Formula: C7H6ClN3
Molecular Weight: 167.60
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 138

Formula: C₇H₆ClN₃
MW: 167.5956 g/mol
Melting Pt: 138 °C
MDL Number: MFCD00216649
CAS Number: 54187-96-1
https://us.vwr.com/stibo/web/std.lang.all/87/48/28938748.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR