Cyclopentylmagnesium bromide ∼18% ∼1 mol/L in THF
C1505-100G
TCC1505-100GEA
135.49
USD
TCC1505-100G
Cyclopentylmagnesium bromide ∼18% ∼1 mol/L in THF
Cyclopentylmagnesium bromide
CAS Number: 33240-34-5
MDL Number: MFCD00209569
Molecular Formula: C5H9BrMg
Molecular Weight: 173.34
Flash Point (°C): -14
Formula:
C₅H₉BrMg Flash Pt: –14 °C |
MDL Number:
MFCD00209569 CAS Number: 33240-34-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...