Cyclohexylmagnesium bromide ∼18% ∼1 mol/L in THF
C1504-100G
TCC1504-100GEA
149.85
USD
TCC1504-100G
Cyclohexylmagnesium bromide ∼18% ∼1 mol/L in THF
Cyclohexylmagnesium bromide
CAS Number: 931-50-0
MDL Number: MFCD01321152
Molecular Formula: C6H11BrMg
Molecular Weight: 187.36
Flash Point (°C): -14
Formula:
C₆H₁₁BrMg Flash Pt: –14 °C |
MDL Number:
MFCD01321152 CAS Number: 931-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...