3-(Carboxymethyl)benzothiazolium bromide ≥97.0% (by titrimetric analysis)

Synonyms: 3-(Carboxymethyl)benzo[d]thiazol-3-ium bromide

C1496-25G
TCC1496-025GEA 129.54 USD
TCC1496-025G
3-(Carboxymethyl)benzothiazolium bromide ≥97.0% (by titrimetric analysis)
3-(Carboxymethyl)benzothiazolium bromide

CAS Number: 74385-09-4
MDL Number: MFCD00134360
Molecular Formula: C9H8BrNO2S
Molecular Weight: 274.13
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Flash Point (°C): 595

Formula: C₉H₈BrNO₂S
MW: 274.1343 g/mol
Flash Pt: 595 °C
MDL Number: MFCD00134360
CAS Number: 74385-09-4
https://us.vwr.com/stibo/web/std.lang.all/87/15/28938715.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR