Cortexolone ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
11-Deoxy-17-hydroxycorticosterone
CAS Number: 152-58-9
MDL Number: MFCD00003662
Molecular Formula: C21H30O4
Molecular Weight: 346.47
Purity/Analysis Method:
Form: Crystal
Specific rotation [a]20/D: 120 deg (C=1, Dioxane)
Product Categories: Bioscience, Pregnanes (Glucocorticoids), Steroids
Other Product Names / Synonyms: 11-Deoxy-17-hydroxycorticosterone, 17alpha,21-Dihydroxy-4-pregnene-3,20-dione, Reichstein's Substance S
Formula:
C₂₁H₃₀O₄ MW: 346.47 g/mol |
MDL Number:
MFCD00003662 CAS Number: 152-58-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...