(Cyanomethyl)tributylphosphonium chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tributyl(cyanomethyl)phosphonium chloride

C1442-10G C1442-25G
TCC1442-25GEA 119.68 USD
TCC1442-25G TCC1442-010G
(Cyanomethyl)tributylphosphonium chloride ≥98.0% (by titrimetric analysis)
(Cyanomethyl)tributylphosphonium chloride

CAS Number: 82358-61-0
MDL Number: MFCD00671508
Molecular Formula: C14H29ClNP
Molecular Weight: 277.82
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 97


Other Product Names / Synonyms: (Cyanomethyl)tributylphosphonium Chloride

Formula: C₁₄H₂₉ClNP
MW: 277.8135 g/mol
Melting Pt: 97 °C
MDL Number: MFCD00671508
CAS Number: 82358-61-0
https://us.vwr.com/stibo/web/std.lang.all/86/67/28938667.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR