N-Cyclohexylformamide ≥98.0%

Supplier: TCI America

Synonyms: N-cyclohexylformamide

C1436-25G
TCC1436-025GEA 60 USD
TCC1436-025G
N-Cyclohexylformamide ≥98.0%
N-Cyclohexylformamide

CAS Number: 766-93-8
MDL Number: MFCD00003828
Molecular Formula: C7H13NO
Molecular Weight: 127.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 151
Flash Point (°C): 113
Freezing point (°C): 30

Formula: C₇H₁₃NO
MW: 127.19 g/mol
Boiling Pt: 151 °C
Flash Pt: 113 °C
MDL Number: MFCD00003828
CAS Number: 766-93-8
https://us.vwr.com/stibo/web/std.lang.all/86/62/28938662.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR