(Chloromethyl)triethoxysilane ≥95.0%
Supplier: TCI America
C1402-10G
C1402-25G
TCC1402-25GEA
69.09
USD
TCC1402-25G
TCC1402-010G
(Chloromethyl)triethoxysilane ≥95.0%
(Chloromethyl)triethoxysilane
CAS Number: 15267-95-5
MDL Number: MFCD00039354
Molecular Formula: C7H17ClO3Si
Molecular Weight: 212.75
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 180
Flash Point (°C): 47
Specific Gravity (20/20): 1.03
Formula:
C₇H₁₇ClO₃Si Boiling Pt: 180 °C Density: 1.03 (20 °C) Flash Pt: 47 °C |
MDL Number:
MFCD00039354 CAS Number: 15267-95-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...