TCIAC1375

C1375-5G
TCC1375-005GEA 52.31 USD
TCC1375-005G
TCIAC1375
CAS Number: 115007-14-2
MDL Number: MFCD00191334
Molecular Formula: C9H16BClF4N2
Molecular Weight: 274.49
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 104

Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis
Other Product Names / Synonyms: Chloro-N,N,N',N'-bis(tetramethylene)formamidinium Tetrafluoroborate, TPyICU
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR