TCIAC1374

C1374-25G
TCC1374-025GEA 67.91 USD
TCC1374-025G
TCIAC1374
CAS Number: 110076-44-3
MDL Number: MFCD00191628
Molecular Formula: C7H8ClNO2S
Molecular Weight: 227.64
Form: Crystal
Color: White

Other Product Names / Synonyms: Sodium o-Toluenesulfonchloramide, o-Toluenesulfonchloramide Sodium Salt
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR