(S)-(-)-2-Chloroisovaleric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (S)-2-Chloro-3-methylbutanoic acid(S)-2-Chloroisovaleric acid(S)-2-Chloro-3-methylbutyric acid

C1365-5G C1365-1G
TCC1365-001GEA 121.75 USD
TCC1365-001G TCC1365-005G
(S)-(-)-2-Chloroisovaleric acid ≥98.0% (by titrimetric analysis)
(S)-(-)-2-Chloroisovaleric acid

CAS Number: 26782-74-1
MDL Number: MFCD00067109
Molecular Formula: C5H9ClO2
Molecular Weight: 136.58
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Boiling point (°C): 109
Specific Gravity (20/20): 1.14
Specific rotation [a]20/D: -1 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: (S)-2-Chloroisovaleric Acid

Formula: C₅H₉ClO₂
MW: 136.58 g/mol
Boiling Pt: 109 °C
Density: 1.14 (20 °C)
MDL Number: MFCD00067109
CAS Number: 26782-74-1
UN: 3265
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/86/04/28938604.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR