4-Nitropyridine-2-carbonitrile ≥99.0%
Supplier: TCI America
Synonyms:
4-Nitro-2-pyridinecarbonitrile
C1359-100MG
TCC1359-100MGEA
18.27
USD
TCC1359-100MG
4-Nitropyridine-2-carbonitrile ≥99.0%
4-Nitropyridine-2-carbonitrile
CAS Number: 19235-88-2
MDL Number: MFCD00191627
Molecular Formula: C6H3N3O2
Molecular Weight: 149.11
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 72
Other Product Names / Synonyms: 4-Nitropicolinonitrile, 4-Nitro-2-pyridinecarbonitrile
Formula:
C₆H₃N₃O₂ MW: 149.1069 g/mol Melting Pt: 72 °C |
MDL Number:
MFCD00191627 CAS Number: 19235-88-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...