Dichloro(chloromethyl)methylsilane ≥97.0%
Supplier: TCI America
CAS Number: 1558-33-4
MDL Number: MFCD00000874
Molecular Formula: C2H5Cl3Si
Molecular Weight: 163.50
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 122
Flash Point (°C): 35
Specific Gravity (20/20): 1.29
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: (Chloromethyl)methyldichlorosilane, Dichloro(chloromethyl)methylsilane
Formula:
C₂H₅Cl₃Si Boiling Pt: 122 °C Density: 1.29 (20 °C) Flash Pt: 35 °C |
MDL Number:
MFCD00000874 CAS Number: 1558-33-4 EINECS: 216-319-5 UN: 2988 ADR: 4.3,I |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...