Decyl chloroformate ≥95.0% (by titrimetric analysis)
Supplier: TCI America
C1357-25ML
TCC1357-025MLEA
148.19
USD
TCC1357-025ML
Decyl chloroformate ≥95.0% (by titrimetric analysis)
Decyl chloroformate
CAS Number: 55488-51-2
MDL Number: MFCD00126853
Molecular Formula: C11H21ClO2
Molecular Weight: 220.74
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 163
Flash Point (°C): 110
Specific Gravity (20/20): 0.96
Other Product Names / Synonyms: Chloroformic Acid Decyl Ester
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...