2-Chloro-4,6-diamino-1,3,5-triazine ≥90.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2,4-Diamino-6-chloro-1,3,5-triazine
, 6-Chloro-1,3,5-triazine-2,4-diamine
C1356-25G
TCC1356-025GEA
128.14
USD
TCC1356-025G
2-Chloro-4,6-diamino-1,3,5-triazine ≥90.0% (by titrimetric analysis)
2-Chloro-4,6-diamino-1,3,5-triazine
CAS Number: 3397-62-4
MDL Number: MFCD00006054
Molecular Formula: C3H4ClN5
Molecular Weight: 145.55
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: 2,4-Diamino-6-chloro-1,3,5-triazine
Formula:
C₃H₄ClN₅ MW: 145.55 g/mol |
MDL Number:
MFCD00006054 CAS Number: 3397-62-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...