1-(2-Chloroethyl)pyrrole ≥98.0%
Supplier: TCI America
Synonyms:
1-(2-Chloroethyl)-1H-pyrrole
C1347-5G
TCC1347-005GEA
318
USD
TCC1347-005G
1-(2-Chloroethyl)pyrrole ≥98.0%
1-(2-Chloroethyl)pyrrole
CAS Number: 77200-24-9
MDL Number: MFCD00191330
Molecular Formula: C6H8ClN
Molecular Weight: 129.59
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 84
Specific Gravity (20/20): 1.13
Formula:
C₆H₈ClN MW: 129.5874 g/mol Boiling Pt: 84 °C Density: 1,13 (20 °C) |
MDL Number:
MFCD00191330 CAS Number: 77200-24-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...