2-Chloro-5-methyl-1,4-benzoquinone ≥98.0%

Supplier: TCI America

Synonyms: 2-Chloro-5-methyl-p-quinone5-Chlorotoluquinone

C1336-5G
TCC1336-005GEA 391.67 USD
TCC1336-005G
2-Chloro-5-methyl-1,4-benzoquinone ≥98.0%
2-Chloro-5-methyl-1,4-benzoquinone

CAS Number: 19832-87-2
MDL Number: MFCD00189381
Molecular Formula: C7H5ClO2
Molecular Weight: 156.57
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 106


Product Categories: Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: 2-Chloro-5-methyl-p-quinone, 5-Chlorotoluquinone

Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Melting Pt: 106 °C
MDL Number: MFCD00189381
CAS Number: 19832-87-2
https://us.vwr.com/stibo/web/std.lang.all/85/78/28938578.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR