2-(Chloromethyl)-1,3-dioxolane ≥95.0%

Supplier: TCI America

Synonyms: 2-Chloromethyl-1,3-dioxolane

C1321-25G
TCC1321-025GEA 28.4 USD
TCC1321-025G
2-(Chloromethyl)-1,3-dioxolane ≥95.0%
2-(Chloromethyl)-1,3-dioxolane

CAS Number: 2568-30-1
MDL Number: MFCD00043402
Molecular Formula: C4H7ClO2
Molecular Weight: 122.55
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Flash Point (°C): 70
Specific Gravity (20/20): 1.25

Formula: C₄H₇ClO₂
MW: 122.55 g/mol
Boiling Pt: 158 °C
Density: 1.25 (20 °C)
Flash Pt: 70 °C
MDL Number: MFCD00043402
CAS Number: 2568-30-1
EINECS: 219-908-5
https://us.vwr.com/stibo/web/std.lang.all/85/63/28938563.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR