1-Cyanomethylpiperidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Piperidinoacetonitrile

C1314-25ML
TCC1314-025MLEA 191.84 USD
TCC1314-025ML
1-Cyanomethylpiperidine ≥98.0% (by GC, titration analysis)
1-Cyanomethylpiperidine

CAS Number: 3010-03-5
MDL Number: MFCD00023733
Molecular Formula: C7H12N2
Molecular Weight: 124.19
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 210
Freezing point (°C): 27


Other Product Names / Synonyms: Piperidinoacetonitrile

Formula: C₇H₁₂N₂
MW: 124.19 g/mol
Boiling Pt: 210 °C
MDL Number: MFCD00023733
CAS Number: 3010-03-5

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR