4-Phenylbenzonitrile ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
p-Phenylbenzonitrile
C1299-5G
C1299-25G
TCC1299-005GEA
36.54
USD
TCC1299-005G
TCC1299-025G
4-Phenylbenzonitrile ≥98.0%
4-Phenylbenzonitrile
CAS Number: 2920-38-9
MDL Number: MFCD00001821
Molecular Formula: C13H9N
Molecular Weight: 179.22
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 193
Melting point (°C): 87
Flash Point (°C): 100
Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Biphenylcarbonitrile, 4-Cyanodiphenyl, 4-Phenylbenzonitrile
Formula:
C₁₃H₉N MW: 179.22 g/mol Boiling Pt: 193 °C Melting Pt: 87 °C Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001821 CAS Number: 2920-38-9 EINECS: 220-860-2 UN: 3439 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...