Cimetidine ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Cyano-1-methyl-3-[2-(5-methyl-1H-imidazol-4-yl-methylthio)ethyl]guanidine

C1252-25G
TCC1252-025GEA 229.42 USD
TCC1252-025G
Cimetidine ≥99.0% (by HPLC, titration analysis)
Cimetidine

CAS Number: 51481-61-9
MDL Number: MFCD00133296
Molecular Formula: C10H16N6S
Molecular Weight: 252.34
Purity/Analysis Method: 99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 141


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 2-Cyano-1-methyl-3-[2-(5-methyl-1H-imidazol-4-yl-methylthio)ethyl]guanidine

Formula: C₁₀H₁₆N₆S
MW: 252.34 g/mol
Melting Pt: 141 °C
MDL Number: MFCD00133296
CAS Number: 51481-61-9

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR