1-(3-Cyanopropyl)pyrrolidine ≥95.0%
Supplier: TCI America
Synonyms:
4-Pyrrolidinobutyronitrile
CAS Number: 35543-25-0
MDL Number: MFCD00003183
Molecular Formula: C8H14N2
Molecular Weight: 138.21
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 99
Specific Gravity (20/20): 0.93
Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: 4-Pyrrolidinobutyronitrile
Formula:
C₈H₁₄N₂ MW: 138.21 g/mol Boiling Pt: 115 °C Density: 0.93 (20 °C) Flash Pt: 99 °C |
MDL Number:
MFCD00003183 CAS Number: 35543-25-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...