1-(4-Chlorophenylsulfonyl)-3-propylurea ≥99.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Chlorpropamide
C1220-25G
TCC1220-025GEA
63.9
USD
TCC1220-025G
1-(4-Chlorophenylsulfonyl)-3-propylurea ≥99.0% (by titrimetric analysis)
1-(4-Chlorophenylsulfonyl)-3-propylurea
CAS Number: 94-20-2
MDL Number: MFCD00079004
Molecular Formula: C10H13ClN2O3S
Molecular Weight: 276.74
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 128
Product Categories: Synthetic Organic Chemistry, Ureas (Multi-component Reaction), Bioscience, Obesity/Diabete Research Reagents
Other Product Names / Synonyms: Chlorpropamide
Formula:
C₁₀H₁₃ClN₂O₃S MW: 276.74 g/mol Melting Pt: 128 °C |
MDL Number:
MFCD00079004 CAS Number: 94-20-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...